Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAGOH
Entrez Gene ID
479562
Gene Name
mago homolog, exon junction complex subunit
Gene Aliases
-
Chromosome Location
Chr.5: 55688443 - 55699741 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_536702.6 XP_536702.1 4 - 5 461 142

View Full Product Details