Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
YBX3
Entrez Gene ID
403728
Gene Name
Y-box binding protein 3
Gene Aliases
CSDA, ZONAB, dpbA
Chromosome Location
Chr.27: 35043756 - 35066230 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF171062.1 - - 458 193

View Full Product Details