Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
CSDC2
Entrez Gene ID
481231
Gene Name
cold shock domain containing C2
Gene Aliases
-
Chromosome Location
Chr.10: 23706962 - 23717078 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_538352.6 XP_538352.4 4 - 4 623 150

View Full Product Details