Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUNDC1
Entrez Gene ID
490955
Gene Name
RUN domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 20071040 - 20080597 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_548078.5 XP_548078.2 - 568 148

View Full Product Details