Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP12
Entrez Gene ID
486033
Gene Name
ubiquitin specific peptidase 12
Gene Aliases
USP12P1
Chromosome Location
Chr.25: 12389360 - 12477406 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022409935.1 XP_022265643.1 - 202 130
XM_543159.6 XP_543159.2 - 302 130

View Full Product Details