Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC71
Entrez Gene ID
608874
Gene Name
coiled-coil domain containing 71
Gene Aliases
-
Chromosome Location
Chr.20: 40031573 - 40036895 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014121981.2 XP_013977456.1 2 - 2 1548 157

View Full Product Details