Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RUNDC1
Entrez Gene ID
490955
Gene Name
RUN domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 20071040 - 20080597 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_548078.5 XP_548078.2 - 1711 107

View Full Product Details