Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
HTR7
Entrez Gene ID
477762
Gene Name
5-hydroxytryptamine receptor 7
Gene Aliases
-
Chromosome Location
Chr.28: 5438872 - 5524385 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005637529.3 XP_005637586.1 2 - 2 813 190
XM_534958.6 XP_534958.2 2 - 2 810 190
GenBank mRNA AJ628248.1 - 2 - 2 575 190
AJ628249.1 - 2 - 2 575 190

View Full Product Details