Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LYSMD3
Entrez Gene ID
488913
Gene Name
LysM domain containing 3
Gene Aliases
-
Chromosome Location
Chr.3: 18412824 - 18425832 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005618120.3 XP_005618177.1 - 593 125
XM_546030.6 XP_546030.2 - 288 125

View Full Product Details