Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PA2G4
Entrez Gene ID
474396
Gene Name
proliferation-associated 2G4
Gene Aliases
EBP1
Chromosome Location
Chr.10: 426074 - 437044 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_531629.5 XP_531629.2 - 1259 84

View Full Product Details