Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRX
Entrez Gene ID
403577
Gene Name
cone-rod homeobox
Gene Aliases
-
Chromosome Location
Chr.1: 108249911 - 108261015 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003049.1 NP_001003049.1 4 - 4 845 153
GenBank mRNA AF454668.1 - 4 - 4 845 153

View Full Product Details