Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCHE
Entrez Gene ID
488143
Gene Name
butyrylcholinesterase
Gene Aliases
-
Chromosome Location
Chr.34: 30912632 - 30977624 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313861.1 NP_001300790.1 2 - 2 1495 118
XM_014110346.2 XP_013965821.1 2 - 2 1536 118

View Full Product Details