Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR6B1
Entrez Gene ID
482725
Gene Name
olfactory receptor family 6 subfamily B member 1
Gene Aliases
-
Chromosome Location
Chr.16: 5884765 - 5885780 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014119839.2 XP_013975314.1 - 224 164

View Full Product Details