Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD13
Entrez Gene ID
479974
Gene Name
sterile alpha motif domain containing 13
Gene Aliases
-
Chromosome Location
Chr.6: 63528376 - 63587419 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005622009.2 XP_005622066.1 2 - 3 147 146
XM_005622010.3 XP_005622067.1 2 - 3 295 146
XM_005622011.3 XP_005622068.1 3 - 4 641 146
XM_005622012.3 XP_005622069.1 2 - 3 545 146
XM_005622013.3 XP_005622070.1 2 - 3 185 146
XM_014114872.2 XP_013970347.1 3 - 4 649 146
XM_014114873.2 XP_013970348.1 3 - 4 659 146
XM_022420580.1 XP_022276288.1 2 - 3 147 146
XM_022420581.1 XP_022276289.1 2 - 3 99 146
XM_022420583.1 XP_022276291.1 3 - 4 651 146
XM_537098.6 XP_537098.4 2 - 3 147 146

View Full Product Details