Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXF1
Entrez Gene ID
489672
Gene Name
forkhead box F1
Gene Aliases
-
Chromosome Location
Chr.5: 66343776 - 66347785 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_546792.6 XP_546792.3 - 536 149

View Full Product Details