Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NMBR
Entrez Gene ID
611644
Gene Name
neuromedin B receptor
Gene Aliases
-
Chromosome Location
Chr.1: 33772687 - 33858112 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022417606.1 XP_022273314.1 - 1961 139
XM_022417607.1 XP_022273315.1 - 1836 139
XM_022417609.1 XP_022273317.1 - 1860 139
XM_022417610.1 XP_022273318.1 - 890 139
XM_022417613.1 XP_022273321.1 - 2928 139

View Full Product Details