Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLDN5
Entrez Gene ID
100684266
Gene Name
claudin 5
Gene Aliases
-
Chromosome Location
Chr.26: 29702464 - 29704950 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005636589.3 XP_005636646.1 1 - 1 1693 102

View Full Product Details