Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
DUSP2
Entrez Gene ID
611653
Gene Name
dual specificity phosphatase 2
Gene Aliases
-
Chromosome Location
Chr.17: 34586635 - 34589076 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005630391.2 XP_005630448.1 3 - 3 603 162

View Full Product Details