Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
CLDN11
Entrez Gene ID
488160
Gene Name
claudin 11
Gene Aliases
-
Chromosome Location
Chr.34: 34912903 - 34929441 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_545284.6 XP_545284.3 3 - 3 574 157

View Full Product Details