Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FICD
Entrez Gene ID
486320
Gene Name
FIC domain protein adenylyltransferase
Gene Aliases
-
Chromosome Location
Chr.26: 18347393 - 18353081 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005636343.3 XP_005636400.1 4 - 4 1206 105
XM_005636344.3 XP_005636401.1 3 - 3 1406 105
XM_022410702.1 XP_022266410.1 3 - 3 1426 105
XM_022410703.1 XP_022266411.1 3 - 3 1179 105
XM_543446.6 XP_543446.3 3 - 3 1123 105

View Full Product Details