Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBIAD1
Entrez Gene ID
487446
Gene Name
UbiA prenyltransferase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.2: 84803338 - 84813030 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544571.4 XP_544571.1 2 - 2 985 125

View Full Product Details