Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SERTAD3
Entrez Gene ID
612504
Gene Name
SERTA domain containing 3
Gene Aliases
-
Chromosome Location
Chr.1: 113258274 - 113264013 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616558.3 XP_005616615.1 3 - 3 570 141
XM_014120362.2 XP_013975837.1 2 - 2 487 141
XM_022425602.1 XP_022281310.1 3 - 3 595 141
XM_022425604.1 XP_022281312.1 2 - 2 493 141
XM_850231.5 XP_855324.2 2 - 2 493 141

View Full Product Details