Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELAC1
Entrez Gene ID
476197
Gene Name
elaC ribonuclease Z 1
Gene Aliases
-
Chromosome Location
Chr.1: 23931360 - 23971516 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022416909.1 XP_022272617.1 - 1022 94

View Full Product Details