Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK10
Entrez Gene ID
490822
Gene Name
potassium two pore domain channel subfamily K member 10
Gene Aliases
-
Chromosome Location
Chr.8: 59484385 - 59612891 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005623724.3 XP_005623781.1 4 - 5 1018 117

View Full Product Details