Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRODH2
Entrez Gene ID
484572
Gene Name
proline dehydrogenase 2
Gene Aliases
-
Chromosome Location
Chr.1: 116831722 - 116842698 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313832.1 NP_001300761.1 3 - 4 601 81

View Full Product Details