Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
KCNK12
Entrez Gene ID
481362
Gene Name
potassium two pore domain channel subfamily K member 12
Gene Aliases
-
Chromosome Location
Chr.10: 49618476 - 49672539 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_538483.6 XP_538483.2 2 - 2 1608 142

View Full Product Details