Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
LTC4S
Entrez Gene ID
607316
Gene Name
leukotriene C4 synthase
Gene Aliases
-
Chromosome Location
Chr.11: 1893927 - 1897028 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_843939.5 XP_849032.3 - 453 88

View Full Product Details