Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD9
Entrez Gene ID
490865
Gene Name
ankyrin repeat domain 9
Gene Aliases
-
Chromosome Location
Chr.8: 70441493 - 70445555 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116161.2 XP_013971636.1 3 - 3 415 97
XM_022422395.1 XP_022278103.1 3 - 3 404 97
XM_547987.6 XP_547987.3 3 - 3 364 97

View Full Product Details