Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NXPH1
Entrez Gene ID
610361
Gene Name
neurexophilin 1
Gene Aliases
-
Chromosome Location
Chr.14: 23953318 - 24250419 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014119202.2 XP_013974677.1 6 - 6 1587 149
XM_847844.5 XP_852937.1 3 - 3 1339 149

View Full Product Details