Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
AQP11
Entrez Gene ID
476798
Gene Name
aquaporin 11
Gene Aliases
-
Chromosome Location
Chr.21: 21170760 - 21185709 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_534003.6 XP_534003.1 2 - 2 974 160

View Full Product Details