The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005641893.3 | XP_005641950.1 | 2 - 2 | 2737 | 120 |
XM_549303.6 | XP_549303.2 | 2 - 2 | 2684 | 120 | |
XM_847854.5 | XP_852947.2 | 2 - 2 | 2476 | 120 |