Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
TTC9C
Entrez Gene ID
483782
Gene Name
tetratricopeptide repeat domain 9C
Gene Aliases
-
Chromosome Location
Chr.18: 53928236 - 53941410 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_540903.6 XP_540903.1 3 - 3 823 150

View Full Product Details