Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LMOD3
Entrez Gene ID
476558
Gene Name
leiomodin 3
Gene Aliases
-
Chromosome Location
Chr.20: 22612916 - 22629207 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005632272.2 XP_005632329.1 - 1314 141
XM_014121536.2 XP_013977011.1 - 1265 141
XM_549501.5 XP_549501.3 - 1242 141

View Full Product Details