Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR20
Entrez Gene ID
611234
Gene Name
WD repeat domain 20
Gene Aliases
-
Chromosome Location
Chr.8: 70150090 - 70208683 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022422389.1 XP_022278097.1 2 - 3 343 109
XM_022422390.1 XP_022278098.1 2 - 3 396 109
XM_022422391.1 XP_022278099.1 2 - 3 206 109
XM_022422392.1 XP_022278100.1 2 - 3 253 109

View Full Product Details