Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNG8
Entrez Gene ID
611498
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 8
Gene Aliases
-
Chromosome Location
Chr.1: 103245102 - 103263154 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616850.3 XP_005616907.1 1 - 2 451 154

View Full Product Details