Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
FAM72A
Entrez Gene ID
607371
Gene Name
family with sequence similarity 72 member A
Gene Aliases
-
Chromosome Location
Chr.38: 2495180 - 2505313 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022415335.1 XP_022271043.1 3 - 3 339 121
XM_844007.4 XP_849100.1 3 - 3 339 121

View Full Product Details