Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
ZFPM2
Entrez Gene ID
481997
Gene Name
zinc finger protein, FOG family member 2
Gene Aliases
-
Chromosome Location
Chr.13: 6435368 - 6796903 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128186.1 NP_001121658.1 - 1003 136
XM_022426499.1 XP_022282207.1 - 1181 136
XM_022426500.1 XP_022282208.1 - 1102 136
XM_022426501.1 XP_022282209.1 - 1065 136
XM_022426502.1 XP_022282210.1 - 976 136
GenBank mRNA EU541502.1 - - 1003 136

View Full Product Details