Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPC2
Entrez Gene ID
403920
Gene Name
NPC intracellular cholesterol transporter 2
Gene Aliases
CE1
Chromosome Location
Chr.8: 47641185 - 47650543 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003242.2 NP_001003242.2 3 - 4 425 73
GenBank mRNA DN392295.1 - 3 - 4 376 73
S77411.1 - 3 - 4 375 73

View Full Product Details