Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLA-64
Entrez Gene ID
474838
Gene Name
MHC class I DLA-64
Gene Aliases
-
Chromosome Location
Chr.12: 983573 - 986969 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014378.1 NP_001014400.1 3 - 4 620 97

View Full Product Details