Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NPC2
- Entrez Gene ID
-
403920
- Gene Name
- NPC intracellular cholesterol transporter 2
- Gene Aliases
-
CE1
- Chromosome Location
-
Chr.8: 47641185 - 47650543 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
S77411.1 |
- |
4 - 4 |
988 |
117 |
View Full Product Details