Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAV2
Entrez Gene ID
475294
Gene Name
caveolin 2
Gene Aliases
CAV-2
Chromosome Location
Chr.14: 55437622 - 55445481 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010997.3 NP_001010997.2 3 - 3 1861 151
XM_005628348.3 XP_005628405.1 2 - 2 1446 151

View Full Product Details