Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK10
Entrez Gene ID
484351
Gene Name
kallikrein related peptidase 10
Gene Aliases
-
Chromosome Location
Chr.1: 105860406 - 105864238 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197158.1 NP_001184087.1 3 - 4 551 86
XM_005615761.3 XP_005615818.1 4 - 5 686 86
GenBank mRNA GQ303578.1 - 3 - 4 551 86

View Full Product Details