Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GUCA2A
Entrez Gene ID
482446
Gene Name
guanylate cyclase activator 2A
Gene Aliases
-
Chromosome Location
Chr.15: 1090109 - 1091277 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198788.1 NP_001185717.1 2 - 3 304 96

View Full Product Details