Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM34
Entrez Gene ID
100568302
Gene Name
tripartite motif containing 34
Gene Aliases
-
Chromosome Location
Chr.21: 28635177 - 28656759 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205191.1 NP_001192120.1 7 - 8 1065 97

View Full Product Details