Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BEX4
Entrez Gene ID
492030
Gene Name
brain expressed X-linked 4
Gene Aliases
BEXL1
Chromosome Location
Chr.X: 76473420 - 76474994 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252047.1 NP_001238976.1 1 - 1 526 97
XM_005641022.3 XP_005641079.1 3 - 3 735 97
GenBank mRNA CO592440.1 - 1 - 1 63 97

View Full Product Details