Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD14
Entrez Gene ID
607076
Gene Name
CD14 molecule
Gene Aliases
-
Chromosome Location
Chr.2: 35790505 - 35792967 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_843653.5 XP_848746.2 - 313 106

View Full Product Details