Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL20
Entrez Gene ID
609203
Gene Name
interleukin 20
Gene Aliases
-
Chromosome Location
Chr.7: 6019879 - 6022707 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022421689.1 XP_022277397.1 - 509 88

View Full Product Details