Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR30A
- Entrez Gene ID
-
100885877
- Gene Name
- microRNA mir-30a
- Gene Aliases
-
cfa-mir-30a
- Chromosome Location
-
Chr.12: 33783894 - 33783956 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_049231.1 |
|
1 - 1 |
36 |
63 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA mir-30c-2 |
MIR30C-2 |
cfa-mir-30c-2 |
-25.8kb
|
Sense |
Chr. - 33757899 - 33757960 on Build CanFam3.1 |
View Full Product Details