Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsc22d2
Entrez Gene ID
100771973
Gene Name
TSC22 domain family, member 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613922: 348333 - 395354 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643492.1 XP_007641682.1 2 - 3 1910 80

View Full Product Details