Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mbtps2
Entrez Gene ID
100689085
Gene Name
membrane-bound transcription factor peptidase, site 2
Gene Aliases
S2P
Chromosome Location
Chr.Un|NW_003613731: 531347 - 575213 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244089.1 NP_001231018.1 6 - 7 865 104
XM_007640118.1 XP_007638308.1 7 - 8 880 104
XM_007640119.1 XP_007638309.1 5 - 6 889 104
GenBank mRNA AF019611.1 - 6 - 7 865 104
JI894193.1 - 1 - 2 122 104
JP050962.1 - 1 - 2 122 104

View Full Product Details